5-Bromonicotinamide ≥98.0% (by GC)
Supplier: TCI EUROPE
|
Warning
|
Synonyms:
5-Bromo-3-pyridinecarboxamide
TCIAB3536-5GEA
72.6
EUR
TCIAB3536-5G
5-Bromonicotinamide ≥98.0% (by GC)
5-Bromonicotinamide
Formula:
C₆H₅BrN₂O MW: 201.03 |
MDL Number:
MFCD00173919 CAS Number: 28733-43-9 |
![]() |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 218.0 to 223.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...