4'-Benzyloxy-2'-hydroxyacetophenone ≥98.0% (by GC)
Supplier: TCI EUROPE
Synonyms:
1-(4-(Benzyloxy)-2-hydroxyphenyl)ethanone
TCIAB3950-1GEA
121
EUR
TCIAB3950-1G
4'-Benzyloxy-2'-hydroxyacetophenone ≥98.0% (by GC)
4'-Benzyloxy-2'-hydroxyacetophenone
Formula:
C₁₅H₁₄O₃ MW: 242.2699 g/mol |
MDL Number:
MFCD00054612 CAS Number: 29682-12-0 |
![]() |
Specification Test Results
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 103.0 to 107.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...