Digital Solutions
Applications
Support
About
Order Entry
Instrument trays, Azlon®
AZLOTWR022
:
SciLabware
Instrument trays, Azlon®
AZLOTWR022
:
SciLabware
365,00 €
1 * 1 ST
-
+
Seq:
03
l×p×h:
610×350×22 mm
Environmentally Preferable:
Cdt:
1
Made of PP, white.
Rectangular
Rigid and robust
Autoclavable