Imprimer
Ubbelohde viscometers (ASTM)
Fournisseur: Fungilab
FUNGCV003-007EA 137 EUR
FUNGCV003-007 FUNGCV003-008 FUNGCV003-016 FUNGCV003-005 FUNGCV003-006 FUNGCV003-009 FUNGCV003-010 FUNGCV003-011 FUNGCV003-014 FUNGCV003-003 FUNGCV003-004 FUNGCV003-015 FUNGCV003-001 FUNGCV003-012 FUNGCV003-013 FUNGCV003-002
Ubbelohde viscometers (ASTM)
Viscosimètres Rhéomètres capillaires
Ubbelohde viscometers are used to determine kinematic viscosity of transparent Newtonian liquids.
Certifications: ASTM
Information de livraison: Supplied with the calibration certificate issued in accordance with the conditions of accreditation granted by a National Metrological Register, the test uncertainty and its traceability to international standards UKAS.