The HM525 NX is a high performance routine cryostat with intuitive software and touch screen. The front of the microtome stops build up of debris behind the microtome making it easier to clean.
Operator safety and minimal service downtime
Cooled specimen positions (27) including four fast-freezing stations to −57 °C to limit ice crystal formation
Additional knee space supports comfortable seated operation
Ten minute specimen fast-freeze function can be accessed from the menu
Optional UV disinfection: High intensity UV light delivers disinfection for peace of mind and enhanced germicidal activity; immediate or scheduled operation; alert screen indicates incomplete or interrupted disinfection cycle and adjustable disinfection time setting up to 300 min.