Disinfectant cleaning wipes, WIP'Anios Excel
DMDA2446655C4EA 119 EUR
DMDA2446655C4
Disinfectant cleaning wipes, WIP'Anios Excel
These 100% biodegradable viscose wipes are ideal for the cleaning and disinfection of external parts of equipment as well as of medical devices which cannot be immersed in liquids. Also suitable for wiping endoscope sheathes.
  • No rinsing required
  • Perfume- and alcohol-free, without CMR substances
  • Broad virucidal spectrum
Composition: Didecyldimethylammonium chloride, polyhexamethylene biguanide hydrochloride, cationic and amphoteric surfactants, chelating agent, excipients.
Microbiological properties: The impregnating solution is bactericidal, fungicidal. Active against M. tuberculosis (Tb), against PRV (Surrogate of HBV), BVDV (Surrogate of HCV), Rotavirus, Herpesvirus, Influenza virus A [H5N1] and [H1N1] and HIV-1.