Imprimer
Pallets and sumps for storage of hazardous materials
Fournisseur: asecos
MSGA23056EA 0 EUR
MSGA23056 MSGA23057 MSGA23058 MSGA23059 MSGA2848 MSGA2847 MSGA23060 MSGA23061
Pallets and sumps for storage of hazardous materials
Stockage des fûts
These pallets and sumps are used for open and streamlined storage box with retention volumes 205 and 400 liters.
  • Flexible wall constructions allow separate storage
  • Stackable on 3 levels
  • Optional swing guard at the front
  • Either painted or completely galvanized