Belgium
Order Entry
Anti-SGK2 Rabbit Polyclonal Antibody
Anti-SGK2 Rabbit Polyclonal Antibody
  ABGEAP7947A
 :  Abgent
undefined
Anti-SGK2 Rabbit Polyclonal Antibody
  ABGEAP7947A
 :  Abgent
 
354,00 €
   
  1 * 400 µl

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Serum/glucocorticoid regulated kinase 2
  • Symbole de l'antigène:
    SGK2
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Cytométrie en flux:
    Yes
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
  • Réactivité:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Épitope:
    N-Terminal (1–100 aa)
  • Adsorption croisée:
    No
  • ID de gène:
    5537
  • Synonymes antigène:
    Serine/threonine-protein kinase Sgk2|Serum/glucocorticoid regulated kinase 2
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequence:
    TREPKLFRAVPDSERC
  • Température de stockage:
    Cool
  • Concentration:
    0,5 mg/ml
  • Température de transport:
    Ice
  • Immunogène:
    This PPP6C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 264-293 amino acids from the C-terminal region of human PPP6C.
  • Purification:
    Protein A & Peptide affinity purification
  • Cdt:
    400 µl

 

 

Type: Primary
Antigen: SGK2 (serum/glucocorticoid regulated kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse