Anti-SGK3 Rabbit Polyclonal Antibody
ABGEAP7949B
: Abgent
- Type d'anticorps:Primaire
- Nom de l'antigène:Serum/glucocorticoid regulated kinase 3
- Symbole de l'antigène:SGK3
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Épitope:Center (123–139 aa)
- Adsorption croisée:No
- ID de gène:30837
- Synonymes antigène:Serine/threonine-protein kinase SGK3|Serum/glucocorticoid regulated kinase-like SGKL|serum/glucocorticoid regulated kinase 3
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:EFIKRAIMHSKNGKC
- Température de stockage:Cool
- Concentration:0,35 mg/ml
- Température de transport:Ice
- Immunogène:This SOCS7 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 458-486 amino acids from the C-terminal region of human SOCS7.
- Purification:Protein A & Peptide affinity purification
- Cdt:400 µl
Type: Primary
Antigen: SGK3 (serum/glucocorticoid regulated kinase family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human