Anti-MAFA Rabbit Polyclonal Antibody
ABGEAP9440C
: Abgent
- Type d'anticorps:Primaire
- Nom de l'antigène:V-maf Musculoaponeurotic Fibrosarcoma Oncogene Homolog A
- Symbole de l'antigène:MAFA
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Cytométrie en flux:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Épitope:N-Terminal
- ID de gène:64114
- Synonymes antigène:hMafA|RIPE3b1
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:SFGPGEWDDRKVRHTC
- Température de stockage:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,5 mg/ml
- Température de transport:4 °C
- Immunogène:This TMBIM1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 68-97 amino acids from the N-terminal region of human TMBIM1.
- Purification:Protein A & Peptide affinity purification
- Cdt:400 µl
Type: Primary
Antigen: MAFA (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse