Anti-PPP1R14A Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3408AEA 350 EUR
ABGEAT3408A
Anti-PPP1R14A Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PPP1R14A (protein phosphatase 1, regulatory (inhibitor) subunit 14A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR