Imprimer

Anti-PPP1R2 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3409AEA 350 EUR
ABGEAT3409A
Anti-PPP1R2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: PPP1R2 (protein phosphatase 1, regulatory (inhibitor) subunit 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR