Anti-PPP2R2B Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3412AEA 350 EUR
ABGEAT3412A
Anti-PPP2R2B Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PPP2R2B (protein phosphatase 2, regulatory subunit B, beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR