Imprimer

Anti-PRKAB1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3428AEA 350 EUR
ABGEAT3428A
Anti-PRKAB1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: PRKAB1 (protein kinase, AMP-activated, beta 1 non-catalytic subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR