Anti-PRPF3 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3442AEA 350 EUR
ABGEAT3442A
Anti-PRPF3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PRPF3 (pre-mRNA processing factor 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR