Anti-TMPRSS15 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3446AEA 350 EUR
ABGEAT3446A
Anti-TMPRSS15 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: TMPRSS15 (transmembrane protease, serine 15)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR