Anti-PSMB6 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3461AEA 350 EUR
ABGEAT3461A
Anti-PSMB6 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PSMB6 (proteasome (prosome, macropain) subunit, beta type, 6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR