Imprimer

Anti-PSMC4 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3466AEA 350 EUR
ABGEAT3466A
Anti-PSMC4 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: PSMC4 (proteasome (prosome, macropain) 26S subunit, ATPase, 4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR