Anti-PSMD5 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3470AEA 350 EUR
ABGEAT3470A
Anti-PSMD5 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PSMD5 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 5)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR