Imprimer

Anti-PSMD7 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3472AEA 350 EUR
ABGEAT3472A
Anti-PSMD7 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: PSMD7 (proteasome (prosome, macropain) 26S subunit, non-ATPase, 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR