Anti-PSPH Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3477AEA 350 EUR
ABGEAT3477A
Anti-PSPH Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PSPH (phosphoserine phosphatase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR