Anti-RAD17 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3544AEA 350 EUR
ABGEAT3544A
Anti-RAD17 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: RAD17 (RAD17 homolog (S. pombe))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR