Anti-RAD51D Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3551AEA 350 EUR
ABGEAT3551A
Anti-RAD51D Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: RAD51D (RAD51 paralog D)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR