Imprimer

Anti-RAPGEFL1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3569AEA 350 EUR
ABGEAT3569A
Anti-RAPGEFL1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RAPGEFL1 (Rap guanine nucleotide exchange factor (GEF)-like 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR