Imprimer

Anti-RARS Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3573AEA 350 EUR
ABGEAT3573A
Anti-RARS Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RARS (arginyl-tRNA synthetase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR