Imprimer

Anti-RASGRP2 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3579AEA 350 EUR
ABGEAT3579A
Anti-RASGRP2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RASGRP2 (RAS guanyl releasing protein 2 (calcium and DAG-regulated))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR