Imprimer

Anti-RASSF8 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3583AEA 350 EUR
ABGEAT3583A
Anti-RASSF8 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RASSF8 (Ras association (RalGDS/AF-6) domain family (N-terminal) member 8)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR