Imprimer

Anti-RDH11 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3609AEA 350 EUR
ABGEAT3609A
Anti-RDH11 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RDH11 (retinol dehydrogenase 11 (all-trans/9-cis/11-cis))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR