Imprimer
Anti-RND2 Mouse Monoclonal Antibody
Fournisseur: Abgent
ABGEAT3655AEA 350 EUR
ABGEAT3655A
Anti-RND2 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RND2 (Rho family GTPase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human