Anti-RNF133 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3664AEA 350 EUR
ABGEAT3664A
Anti-RNF133 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: RNF133 (ring finger protein 133)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR