Imprimer

Anti-MARCH10 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3671AEA 350 EUR
ABGEAT3671A
Anti-MARCH10 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RNF190
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR