Imprimer

Anti-RRP9 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3682AEA 350 EUR
ABGEAT3682A
Anti-RRP9 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RRP9 (ribosomal RNA processing 9, small subunit (SSU) processome component, homolog (yeast))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR