Imprimer
Anti-ROPN1 Mouse Monoclonal Antibody
Fournisseur: Abgent
ABGEAT3690AEA 350 EUR
ABGEAT3690A
Anti-ROPN1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: ROPN1 (rhophilin associated tail protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human