Imprimer

Anti-RPL39L Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3708AEA 350 EUR
ABGEAT3708A
Anti-RPL39L Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RPL39L (ribosomal protein L39-like)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR