Imprimer

Anti-RPL9 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3710AEA 350 EUR
ABGEAT3710A
Anti-RPL9 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RPL9 (ribosomal protein L9)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR