Anti-RPS6KB1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3724AEA 350 EUR
ABGEAT3724A
Anti-RPS6KB1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: RPS6KB1 (ribosomal protein S6 kinase, 70kDa, polypeptide 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR