Anti-RUFY1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3735AEA 350 EUR
ABGEAT3735A
Anti-RUFY1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: RUFY1 (RUN and FYVE domain containing 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR