Imprimer

Anti-RXRB Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3748AEA 350 EUR
ABGEAT3748A
Anti-RXRB Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: RXRB (retinoid X receptor, beta)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR