Imprimer

Anti-S100A6 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3760AEA 350 EUR
ABGEAT3760A
Anti-S100A6 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: S100A6 (S100 calcium binding protein A6)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR