Imprimer

Anti-SAFB Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3767AEA 350 EUR
ABGEAT3767A
Anti-SAFB Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SAFB (scaffold attachment factor B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR