Anti-SCMH1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3788AEA 350 EUR
ABGEAT3788A
Anti-SCMH1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SCMH1 (sex comb on midleg homolog 1 (Drosophila))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR