Anti-AIMP1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3796AEA 350 EUR
ABGEAT3796A
Anti-AIMP1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: AIMP1 (aminoacyl tRNA synthetase complex-interacting multifunctional protein 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR