Anti-SDHC Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3802AEA 350 EUR
ABGEAT3802A
Anti-SDHC Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SDHC (succinate dehydrogenase complex, subunit C, integral membrane protein, 15kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR