Imprimer

Anti-SEMA4D Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3808AEA 350 EUR
ABGEAT3808A
Anti-SEMA4D Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SEMA4D (sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR