Imprimer

Anti-SERPINA12 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3817AEA 350 EUR
ABGEAT3817A
Anti-SERPINA12 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SERPINA12 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 12)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR