Anti-SERPINE2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3825AEA 350 EUR
ABGEAT3825A
Anti-SERPINE2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SERPINE2 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR