Imprimer

Anti-SERPINH1 Mouse Monoclonal Antibody

Fournisseur: Abgent
ABGEAT3827AEA 350 EUR
ABGEAT3827A
Anti-SERPINH1 Mouse Monoclonal Antibody
Anticorps
Type: Primary
Antigen: SERPINH1 (serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR