Anti-SH3BP5 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3871AEA 350 EUR
ABGEAT3871A
Anti-SH3BP5 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SH3BP5 (SH3-domain binding protein 5 (BTK-associated))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR