Anti-SHC2 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3881AEA 350 EUR
ABGEAT3881A
Anti-SHC2 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SHC2 (SHC (Src homology 2 domain containing) transforming protein 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR