Anti-SLC15A1 Mouse Monoclonal Antibody

Supplier: Abgent
ABGEAT3902AEA 350 EUR
ABGEAT3902A
Anti-SLC15A1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: SLC15A1 (solute carrier family 15 (oligopeptide transporter), member 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR